siRNA Details
| Gene Symbol: Slc4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009208.3 | NP_033234.2 | 19, 20 | 3228 | ||
| GenBank mRNA | AK147218.1 | - | 3565 | |||
| BC066086.1 | - | 20, 21 | 3102 | |||
| BC080271.1 | - | 19, 20 | 3223 | 6831942 | ||
| M28383.1 | - | 19, 20 | 3198 | |||

Slc4a3
Genomic Map