siRNA Details
| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011404.3 | NP_035534.2 | 8 | 1343 | ||
| GenBank mRNA | AB017189.1 | - | 8 | 1289 | ||
| AB023409.1 | - | 8 | 1271 | |||
| AK142288.1 | - | 8 | 1344 | |||
| AK148134.1 | - | 8 | 1013 | |||
| AK178111.1 | - | 1 | 110 | |||
| AK186391.1 | - | 1 | 110 | |||
| AK187399.1 | - | 1 | 111 | |||
| AK190487.1 | - | 1 | 110 | |||
| AK190616.1 | - | 1 | 110 | |||
| AK191108.1 | - | 1 | 110 | |||
| AK191383.1 | - | 1 | 110 | |||
| AK192542.1 | - | 1 | 110 | |||
| AK193936.1 | - | 1 | 110 | |||
| AK195990.1 | - | 1 | 110 | |||
| AK196017.1 | - | 1 | 110 | |||
| AK196254.1 | - | 1 | 110 | |||
| AK196383.1 | - | 1 | 110 | |||
| AK197058.1 | - | 1 | 111 | |||
| AK197141.1 | - | 1 | 110 | |||
| AK197882.1 | - | 1 | 110 | |||
| AK198482.1 | - | 1 | 110 | |||
| AK198655.1 | - | 1 | 110 | |||
| AK198981.1 | - | 1 | 110 | |||
| AK199144.1 | - | 1 | 110 | |||
| AK199202.1 | - | 1 | 110 | |||
| AK199418.1 | - | 1 | 110 | |||
| AK199753.1 | - | 1 | 110 | |||
| AK200076.1 | - | 1 | 110 | |||
| AK200375.1 | - | 1 | 110 | |||
| AK201908.1 | - | 1 | 110 | |||
| AK201954.1 | - | 1 | 110 | |||
| AK204088.1 | - | 1 | 111 | |||
| AK204407.1 | - | 1 | 115 | |||
| AK205035.1 | - | 1 | 110 | |||
| AK207173.1 | - | 1 | 117 | |||
| AK217749.1 | - | 1 | 104 | |||
| BC026131.1 | - | 8 | 1352 | 4188854 | ||

Slc7a5
Genomic Map