siRNA Details
| Gene Symbol: Slfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF099976.1 | - | 2 | 860 | ||
| AK137559.1 | - | 8 | 2505 | |||
| AK137689.1 | - | 7 | 2426 | |||
| BC044865.1 | - | 8 | 2514 | 4459564 | ||
| BC046511.1 | - | 3 | 393 | |||
| BC150770.1 | - | 5 | 2121 | 9087681 | ||

Slfn4
Genomic Map