siRNA Details
| Gene Symbol: Slit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015748.3 | NP_056563.2 | 9, 10 | 1400 | ||
| GenBank mRNA | AF144627.1 | - | 9, 10 | 1425 | ||
| AK122376.1 | - | 9, 10 | 1166 | |||
| BC057131.1 | - | 9, 10 | 1206 | 6827560 | ||
| BC062091.1 | - | 9, 10 | 1206 | 6827560 | ||

Slit1
Genomic Map