siRNA Details
| Gene Symbol: Slit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011412.3 | NP_035542.2 | 14, 15 | 1528 | ||
| GenBank mRNA | AF144629.1 | - | 14, 15 | 1469 | ||
| AK147292.1 | - | 14, 15 | 1529 | |||
| BC150780.1 | - | 14, 15 | 1505 | 9087691 | ||

Slit3
Genomic Map