siRNA Details
| Gene Symbol: Hltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009210.3 | NP_033236.2 | 10 | 1251 | ||
| GenBank mRNA | AB221636.1 | - | 10 | 1074 | ||
| AF010600.1 | - | 10 | 1251 | |||
| AF165911.1 | - | 10 | 1230 | |||
| AK141235.1 | - | 10 | 1244 | |||
| AK142961.1 | - | 10 | 1208 | |||
| AK149897.1 | - | 10 | 1289 | |||
| AK166911.1 | - | 10 | 1221 | |||
| BC057116.1 | - | 10 | 1220 | 6408672 | ||
| BC059240.1 | - | 10 | 1220 | 6408672 | ||

Hltf
Genomic Map