siRNA Details
| Gene Symbol: Kdm5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013668.4 | NP_038696.2 | 4 | 805 | ||
| GenBank mRNA | AF127245.1 | - | 4 | 760 | ||
| AK008105.1 | - | 4 | 790 | |||
| AK011577.1 | - | 4 | 794 | |||
| AK081369.1 | - | 4 | 779 | |||
| AK150152.1 | - | 4 | 805 | |||
| AK155279.1 | - | 4 | 790 | |||
| AK155427.1 | - | 4 | 790 | |||
| AK155651.1 | - | 4 | 803 | |||
| AK158340.1 | - | 4 | 790 | |||
| BC026777.1 | - | 4 | 776 | |||
| BC038940.1 | - | 4 | 791 | |||
| BC043096.1 | - | 3 | 637 | 6405943 | ||
| BC054550.1 | - | 3 | 637 | 6405943 | ||
| Z29651.1 | - | 4 | 367 | |||

Kdm5c
Genomic Map