siRNA Details
| Gene Symbol: Kdm5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013668.4 | NP_038696.2 | 25 | 4545 | ||
| GenBank mRNA | AF127245.1 | - | 25 | 4500 | ||
| AK129096.1 | - | 22 | 3728 | |||
| AK155651.1 | - | 25 | 4543 | |||
| BC043096.1 | - | 24 | 4377 | 6405943 | ||
| BC054550.1 | - | 24 | 4377 | 6405943 | ||

Kdm5c
Genomic Map