siRNA Details
| Gene Symbol: Kdm5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013668.4 | NP_038696.2 | 17 | 2714 | ||
| GenBank mRNA | AF127245.1 | - | 17 | 2669 | ||
| AK129096.1 | - | 14 | 1888 | |||
| AK155651.1 | - | 17 | 2712 | |||
| BC043096.1 | - | 16 | 2546 | 6405943 | ||
| BC054550.1 | - | 16 | 2546 | 6405943 | ||
| Z29651.1 | - | 17 | 2276 | |||

Kdm5c
Genomic Map