siRNA Details
| Gene Symbol: Snrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164572.1 | NP_001158044.1 | 4 | 1076 | ||
| NM_133741.2 | NP_598502.2 | 5 | 1126 | |||
| GenBank mRNA | AF387809.2 | - | 4 | 975 | ||
| AK082185.1 | - | 1737 | ||||
| AK182697.1 | - | 349 | ||||
| AK189350.1 | - | 3 | 304 | |||
| BC020189.1 | - | 5 | 1017 | 4482701 | ||
| BC094658.1 | - | 4 | 1003 | 5721174 | ||
| BC150742.1 | - | 3 | 947 | 9087653 | ||

Snrk
Genomic Map