siRNA Details
| Gene Symbol: Snrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164572.1 | NP_001158044.1 | 6 | 1745 | ||
| NM_133741.2 | NP_598502.2 | 7 | 1795 | |||
| GenBank mRNA | AF387809.2 | - | 6 | 1644 | ||
| BC020189.1 | - | 7 | 1686 | 4482701 | ||
| BC094658.1 | - | 6 | 1672 | 5721174 | ||
| BC116410.1 | - | 1 | 175 | |||
| BC150742.1 | - | 5 | 1616 | 9087653 | ||

Snrk
Genomic Map