siRNA Details
| Gene Symbol: Sort1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271599.1 | NP_001258528.1 | 4, 5 | 610 | ||
| NM_019972.3 | NP_064356.2 | 4, 5 | 610 | |||
| GenBank mRNA | AF175279.1 | - | 4, 5 | 539 | ||
| AK147442.1 | - | 4, 5 | 593 | |||
| BC034129.1 | - | 1, 2 | 43 | |||
| BC056343.1 | - | 4, 5 | 542 | 5704346 | ||

Sort1
Genomic Map