siRNA Details
Gene Symbol: Sos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009231.2 | NP_033257.2 | 10 | 1867 | ||
GenBank mRNA | AK140310.1 | - | 10 | 1867 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009231.2 | NP_033257.2 | 10 | 1867 | ||
GenBank mRNA | AK140310.1 | - | 10 | 1867 |