siRNA Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113559.2 | NP_001107031.2 | 13 | 1967 | ||
| NM_001243163.1 | NP_001230092.1 | 13 | 1859 | |||
| NM_011444.3 | NP_035574.2 | 14 | 2111 | |||
| GenBank mRNA | AB006330.1 | - | 14 | 2023 | ||
| AJ010604.1 | - | 13 | 1849 | |||
| AK160154.1 | - | 7 | 925 | |||
| BC110478.2 | - | 13 | 1851 | 40047865 | ||
| KF793916.1 | - | 14 | 1812 | |||
| X65657.1 | - | 7 | 896 | |||

Sox5
Genomic Map