siRNA Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113559.2 | NP_001107031.2 | 12 | 1667 | ||
| NM_001243163.1 | NP_001230092.1 | 12 | 1559 | |||
| NM_011444.3 | NP_035574.2 | 13 | 1811 | |||
| GenBank mRNA | AB006330.1 | - | 13 | 1723 | ||
| AJ010604.1 | - | 12 | 1549 | |||
| AK015212.1 | - | 6 | 620 | |||
| AK160154.1 | - | 6 | 625 | |||
| BC110478.2 | - | 12 | 1551 | 40047865 | ||
| KF793916.1 | - | 13 | 1512 | |||
| X65657.1 | - | 6 | 596 | |||

Sox5
Genomic Map