siRNA Details
| Gene Symbol: Sp100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013673.3 | NP_038701.2 | 14, 15 | 1277 | ||
| GenBank mRNA | AK083304.1 | - | 14, 15 | 1278 | ||
| AK152348.1 | - | 13, 14 | 1260 | |||
| AK153487.1 | - | 12, 13 | 1185 | |||
| BC069183.1 | - | 13, 14 | 1246 | 4911025 | ||
| CT010199.1 | - | 14, 15 | 1225 | |||
| U83630.1 | - | 14, 15 | 1314 | |||
| U83636.1 | - | 13, 14 | 1171 | |||

Sp100
Genomic Map