siRNA Details
| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018042.3 | NP_001018052.1 | 7 | 2243 | ||
| NM_001098425.1 | NP_001091895.1 | 5 | 2152 | |||
| GenBank mRNA | AF062567.1 | - | 5 | 1879 | ||
| AK004607.1 | - | 4 | 1893 | |||
| AK088238.1 | - | 5 | 2127 | |||
| AK132702.1 | - | 5 | 1898 | |||
| BC027797.1 | - | 4 | 1365 | |||
| BC079874.1 | - | 7 | 2243 | 6833706 | ||

Sp3
Genomic Map