siRNA Details
| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018042.3 | NP_001018052.1 | 4 | 1409 | ||
| NM_001098425.1 | NP_001091895.1 | 2 | 1318 | |||
| GenBank mRNA | AF062567.1 | - | 2 | 1045 | ||
| AK004607.1 | - | 1 | 1059 | |||
| AK088238.1 | - | 2 | 1293 | |||
| AK132702.1 | - | 2 | 1064 | |||
| BC027797.1 | - | 1 | 531 | |||
| BC079874.1 | - | 4 | 1409 | 6833706 | ||

Sp3
Genomic Map