siRNA Details
| Gene Symbol: Sp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166385.2 | NP_001159857.2 | 3 | 914 | ||
| NM_009239.4 | NP_033265.4 | 3 | 914 | |||
| GenBank mRNA | AB221633.1 | - | 2 | 707 | ||
| BC076630.1 | - | 3 | 844 | 30637762 | ||
| U62522.1 | - | 3 | 843 | |||

Sp4
Genomic Map