siRNA Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199012.2 | NP_950177.2 | 11, 12 | 1238 | ||
| GenBank mRNA | AK122368.1 | - | 11, 12 | 1130 | ||
| BC060647.1 | - | 11, 12 | 1200 | 6854041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fchsd2| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199012.2 | NP_950177.2 | 11, 12 | 1238 | ||
| GenBank mRNA | AK122368.1 | - | 11, 12 | 1130 | ||
| BC060647.1 | - | 11, 12 | 1200 | 6854041 | ||