siRNA Details
| Gene Symbol: Spic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011461.3 | NP_035591.3 | 6 | 810 | ||
| GenBank mRNA | AF098863.2 | - | 6 | 809 | ||
| AK137522.1 | - | 6 | 813 | |||
| AK156024.1 | - | 6 | 811 | |||
| AK156892.1 | - | 6 | 811 | |||
| AK157188.1 | - | 7 | 905 | |||
| BC064038.1 | - | 5 | 771 | 3824951 | ||

Spic
Genomic Map