siRNA Details
Gene Symbol: Fam120c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198105.2 | NP_932773.2 | 10 | 2098 | ||
GenBank mRNA | AK030477.1 | - | 10 | 2147 | ||
AK136727.1 | - | 10 | 2094 | |||
AK156920.1 | - | 10 | 2099 | |||
AY150023.1 | - | 10 | 2102 | |||
AY150024.1 | - | 10 | 2102 | |||
BC137973.1 | - | 10 | 2098 | 8860992 | ||
BC171940.1 | - | 10 | 2098 | 9054226 |
