siRNA Details
| Gene Symbol: Fam120c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198105.2 | NP_932773.2 | 13, 14 | 2850 | ||
| GenBank mRNA | AK030477.1 | - | 13, 14 | 2899 | ||
| AK136727.1 | - | 13, 14 | 2846 | |||
| AK156920.1 | - | 13, 14 | 2851 | |||
| AY150023.1 | - | 13, 14 | 2854 | |||
| AY150024.1 | - | 13, 14 | 2854 | |||
| BC137973.1 | - | 13, 14 | 2850 | 8860992 | ||
| BC171940.1 | - | 13, 14 | 2850 | 9054226 | ||

Fam120c
Genomic Map