siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 11 | 1367 | ||
| GenBank mRNA | AK078956.1 | - | 2 | 158 | ||
| BC014710.1 | - | 12 | 1451 | 4502840 | ||
| X84692.1 | - | 1304 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strbp| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009261.3 | NP_033287.2 | 11 | 1367 | ||
| GenBank mRNA | AK078956.1 | - | 2 | 158 | ||
| BC014710.1 | - | 12 | 1451 | 4502840 | ||
| X84692.1 | - | 1304 | ||||