siRNA Details
| Gene Symbol: Kctd12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175429.3 | NP_780638.1 | 3 | 651 | ||
| GenBank mRNA | AK046557.1 | - | 3 | 540 | ||
| AK046708.1 | - | 3 | 578 | |||
| AK050097.1 | - | 2 | 513 | |||
| AK085035.1 | - | 3 | 579 | |||
| BC064459.1 | - | 3 | 651 | 6821736 | ||

Kctd12b
Genomic Map