siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 3 | 692 | ||
| GenBank mRNA | AK004941.1 | - | 3 | 502 | ||
| AK154720.1 | - | 696 | ||||
| AK207238.1 | - | 2 | 97 | |||
| AK216452.1 | - | 2 | 97 | |||
| BC010400.1 | - | 3 | 689 | 4219245 | ||
| BC054423.1 | - | 3 | 676 | 4985039 | ||
| S71375.1 | - | 1 | 51 | |||
| S77493.1 | - | 3 | 694 | |||
| W65601.1 | - | 2 | 151 | |||
| Z21947.1 | - | 1 | 51 | |||

Spr
Genomic Map