siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 3 | 662 | ||
| GenBank mRNA | AK004941.1 | - | 3 | 472 | ||
| AK154720.1 | - | 666 | ||||
| AK207238.1 | - | 2 | 67 | |||
| AK216452.1 | - | 2 | 67 | |||
| BC010400.1 | - | 3 | 659 | 4219245 | ||
| BC054423.1 | - | 3 | 646 | 4985039 | ||
| S71375.1 | - | 1 | 21 | |||
| S77493.1 | - | 3 | 664 | |||
| W65601.1 | - | 2 | 121 | |||
| Z21947.1 | - | 1 | 21 | |||

Spr
Genomic Map