siRNA Details
| Gene Symbol: Camkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199676.1 | NP_001186605.1 | 15 | 1681 | ||
| NM_145358.2 | NP_663333.1 | 15 | 1681 | |||
| GenBank mRNA | AF453383.1 | - | 14 | 1464 | ||
| AK031399.1 | - | 15 | 1606 | |||
| AK032070.1 | - | 15 | 1654 | |||
| AK122370.1 | - | 10 | 847 | |||
| BC023103.1 | - | 14 | 1514 | 3993474 | ||
| BC025895.1 | - | 3 | 135 | |||
| CK625345.1 | - | 3 | 211 | |||

Camkk2
Genomic Map