siRNA Details
| Gene Symbol: Tbc1d16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172443.3 | NP_766031.2 | 10, 11 | 2073 | ||
| GenBank mRNA | AK047555.1 | - | 10, 11 | 2074 | ||
| BC057634.1 | - | 10, 11 | 2032 | 2866041 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d16| Gene Symbol: Tbc1d16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172443.3 | NP_766031.2 | 10, 11 | 2073 | ||
| GenBank mRNA | AK047555.1 | - | 10, 11 | 2074 | ||
| BC057634.1 | - | 10, 11 | 2032 | 2866041 | ||