siRNA Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174847.2 | NP_777272.2 | 11 | 1649 | ||
| GenBank mRNA | AK017896.1 | - | 11 | 1634 | ||
| AK077731.1 | - | 11 | 1610 | |||
| AK080956.1 | - | 11 | 1593 | |||
| AK086520.1 | - | 11 | 1652 | |||
| AK143875.1 | - | 11 | 1623 | |||
| AK169902.1 | - | 11 | 1585 | |||
| AK220454.1 | - | 11 | 1634 | |||
| BC038064.1 | - | 1 | 81 | |||
| BC043688.1 | - | 11 | 1610 | 5136661 | ||

C2cd2
Genomic Map