siRNA Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174847.2 | NP_777272.2 | 13 | 2074 | ||
| GenBank mRNA | AF416778.1 | - | 1 | 41 | ||
| AK077731.1 | - | 13 | 2035 | |||
| AK080956.1 | - | 13 | 2018 | |||
| AK086520.1 | - | 13 | 2077 | |||
| AK220454.1 | - | 13 | 2059 | |||
| BC038064.1 | - | 3 | 506 | |||
| BC043688.1 | - | 13 | 2035 | 5136661 | ||

C2cd2
Genomic Map