siRNA Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174847.2 | NP_777272.2 | 14 | 2394 | ||
| GenBank mRNA | AK077731.1 | - | 14 | 2355 | ||
| AK080956.1 | - | 14 | 2338 | |||
| AK086520.1 | - | 14 | 2397 | |||
| AK220454.1 | - | 14 | 2379 | |||
| BC038064.1 | - | 4 | 826 | |||
| BC043688.1 | - | 14 | 2355 | 5136661 | ||

C2cd2
Genomic Map