siRNA Details
| Gene Symbol: Gm608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029889.2 | NP_001025060.1 | 8 | 2910 | ||
| GenBank mRNA | BC141330.1 | - | 6 | 2699 | 9055942 | |
| BC141331.1 | - | 6 | 2699 | 9055943 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm608| Gene Symbol: Gm608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029889.2 | NP_001025060.1 | 8 | 2910 | ||
| GenBank mRNA | BC141330.1 | - | 6 | 2699 | 9055942 | |
| BC141331.1 | - | 6 | 2699 | 9055943 | ||