siRNA Details
| Gene Symbol: 4930539E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172450.3 | NP_766038.1 | 11 | 2020 | ||
| GenBank mRNA | AK019726.1 | - | 210 | |||
| AK033585.1 | - | 11 | 2021 | |||
| BC117930.2 | - | 11 | 2016 | 40097681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930539E08Rik| Gene Symbol: 4930539E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172450.3 | NP_766038.1 | 11 | 2020 | ||
| GenBank mRNA | AK019726.1 | - | 210 | |||
| AK033585.1 | - | 11 | 2021 | |||
| BC117930.2 | - | 11 | 2016 | 40097681 | ||