siRNA Details
| Gene Symbol: Mchr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145132.2 | NP_660114.1 | 2 | 671 | ||
| GenBank mRNA | AF498247.1 | - | 2 | 396 | ||
| AF498248.1 | - | 2 | 396 | |||
| AK134867.1 | - | 2 | 554 | |||
| BC128285.1 | - | 2 | 394 | |||
| BC128286.1 | - | 2 | 396 | 40102363 | ||
More Information
Set Membership: |
|

Mchr1
Genomic Map