siRNA Details
| Gene Symbol: Esrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290383.1 | NP_001277312.1 | 10 | 1428 | ||
| NM_194055.3 | NP_918944.2 | 10 | 1431 | |||
| GenBank mRNA | AK042164.1 | - | 10 | 1043 | ||
| AK140848.1 | - | 10 | 1429 | |||
| BC031468.1 | - | 8 | 734 | 4037840 | ||
| BC059280.1 | - | 8 | 732 | |||
| DV654302.1 | - | 5 | 492 | |||

Esrp1
Genomic Map