siRNA Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029838.2 | NP_001025009.1 | 13 | 1555 | ||
| NM_148950.3 | NP_683752.2 | 12 | 1450 | |||
| GenBank mRNA | AB086817.1 | - | 10 | 1276 | ||
| AF487460.1 | - | 11 | 1356 | |||
| AK082952.1 | - | 13 | 1500 | |||
| BC050865.1 | - | 12 | 1384 | 6332968 | ||

Pknox2
Genomic Map