siRNA Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029838.2 | NP_001025009.1 | 7 | 721 | ||
| NM_148950.3 | NP_683752.2 | 6 | 616 | |||
| GenBank mRNA | AB086817.1 | - | 4 | 442 | ||
| AF487460.1 | - | 5 | 522 | |||
| AK076428.1 | - | 1 | 1352 | |||
| AK082952.1 | - | 7 | 666 | |||
| AK084178.1 | - | 7 | 664 | |||
| BC050865.1 | - | 6 | 550 | 6332968 | ||

Pknox2
Genomic Map