siRNA Details
| Gene Symbol: Tmprss7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172455.3 | NP_766043.3 | 14 | 1642 | ||
| GenBank mRNA | AK045663.1 | - | 12 | 1392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss7| Gene Symbol: Tmprss7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172455.3 | NP_766043.3 | 14 | 1642 | ||
| GenBank mRNA | AK045663.1 | - | 12 | 1392 | ||