siRNA Details
| Gene Symbol: Btbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145361.2 | NP_663336.2 | 2, 3 | 1064 | ||
| GenBank mRNA | BC055704.1 | - | 1, 2 | 131 | ||
| BC083320.1 | - | 1, 2 | 37 | |||
| BC115682.1 | - | 1, 2 | 124 | 40104217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btbd2| Gene Symbol: Btbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145361.2 | NP_663336.2 | 2, 3 | 1064 | ||
| GenBank mRNA | BC055704.1 | - | 1, 2 | 131 | ||
| BC083320.1 | - | 1, 2 | 37 | |||
| BC115682.1 | - | 1, 2 | 124 | 40104217 | ||