siRNA Details
| Gene Symbol: Dip2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081426.2 | NP_001074895.2 | 37 | 4950 | ||
| GenBank mRNA | AK036188.1 | - | 16 | 1805 | ||
| AK173066.1 | - | 29 | 3362 | |||
| BC056940.1 | - | 20 | 2414 | |||
| BC066797.1 | - | 17 | 1996 | |||
| BC089303.1 | - | 17 | 1974 | |||
| BC137787.1 | - | 37 | 4542 | 8860806 | ||
| BC144909.1 | - | 36 | 4455 | 9053457 | ||
| BC144910.1 | - | 37 | 4542 | 9053458 | ||

Dip2c
Genomic Map