siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 2, 3 | 225 | ||
| GenBank mRNA | AK014431.1 | - | 2, 3 | 234 | ||
| AK019725.2 | - | 2, 3 | 265 | |||
| AK054470.1 | - | 2, 3 | 225 | |||
| AK136327.1 | - | 2, 3 | 235 | |||
| BC060970.1 | - | 2, 3 | 236 | |||
| BC082283.1 | - | 2, 3 | 236 | 30436512 | ||
| L36062.1 | - | 2, 3 | 241 | |||

Star
Genomic Map