siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 4 | 494 | ||
| GenBank mRNA | AK019725.2 | - | 4 | 534 | ||
| AK054470.1 | - | 4 | 494 | |||
| AK136327.1 | - | 4 | 504 | |||
| BC082283.1 | - | 4 | 505 | 30436512 | ||
| L36062.1 | - | 4 | 510 | |||

Star
Genomic Map