siRNA Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205314.1 | NP_001192243.1 | 16, 17 | 1680 | ||
| NM_009283.4 | NP_033309.3 | 16, 17 | 1695 | |||
| GenBank mRNA | AK018544.1 | - | 16, 17 | 1646 | ||
| AK039458.1 | - | 16, 17 | 1696 | |||
| AK082706.1 | - | 16, 17 | 1677 | |||
| AK084855.1 | - | 16, 17 | 1629 | |||
| AK157263.1 | - | 16, 17 | 1544 | |||
| AK159887.1 | - | 16, 17 | 1679 | |||
| BC004808.1 | - | 16, 17 | 1631 | 3587831 | ||
| U06924.1 | - | 13, 14 | 1349 | |||

Stat1
Genomic Map