siRNA Details
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019963.1 | NP_064347.1 | 4 | 469 | ||
GenBank mRNA | AF088862.1 | - | 4 | 339 | ||
AF187231.1 | - | 4 | 463 | |||
AF206162.1 | - | 4 | 469 | |||
AK149838.1 | - | 4 | 466 | |||
AK149924.1 | - | 4 | 482 | |||
AK150070.1 | - | 4 | 481 | |||
AK153405.1 | - | 2 | 107 |
