siRNA Details
| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011486.4 | NP_035616.1 | 23, 24 | 2486 | ||
| NM_213659.2 | NP_998824.1 | 23, 24 | 2536 | |||
| NM_213660.2 | NP_998825.1 | 23, 24 | 2533 | |||
| GenBank mRNA | AK079406.1 | - | 109 | |||
| AK145486.1 | - | 23, 24 | 2392 | |||
| AK153005.1 | - | 23, 24 | 2448 | |||
| AK153472.1 | - | 23, 24 | 2448 | |||
| AY299489.1 | - | 22, 23 | 2263 | |||
| AY299490.1 | - | 22, 23 | 2263 | |||
| BC003806.1 | - | 23, 24 | 2424 | 3593588 | ||
| BC019168.1 | - | 23, 24 | 2390 | 4923137 | ||
| BC037688.1 | - | 7, 8 | 842 | |||
| L29278.1 | - | 23, 24 | 2515 | |||
| U06922.1 | - | 23, 24 | 2325 | |||
| U08378.1 | - | 23, 24 | 2400 | |||
| U30709.1 | - | 23, 24 | 2379 | |||

Stat3
Genomic Map