siRNA Details
| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088991.1 | - | 5 | 448 | ||
| AK132675.1 | - | 22 | 2091 | |||
| BC052907.1 | - | 21 | 1971 | |||
| BC098499.1 | - | 21 | 1958 | 30074608 | ||
| U06923.1 | - | 21 | 1964 | |||
| U09351.1 | - | 21 | 2415 | |||

Stat4
Genomic Map