siRNA Details
| Gene Symbol: Kntc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042421.1 | NP_001035886.1 | 47 | 4996 | ||
| GenBank mRNA | AK084529.1 | - | 4 | 403 | ||
| AK172905.1 | - | 47 | 4970 | |||
| BC023686.1 | - | 14 | 1726 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kntc1| Gene Symbol: Kntc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042421.1 | NP_001035886.1 | 47 | 4996 | ||
| GenBank mRNA | AK084529.1 | - | 4 | 403 | ||
| AK172905.1 | - | 47 | 4970 | |||
| BC023686.1 | - | 14 | 1726 | |||