siRNA Details
| Gene Symbol: Tspan10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145363.2 | NP_663338.2 | 3 | 950 | ||
| GenBank mRNA | AK084214.1 | - | 3 | 897 | ||
| AK148037.1 | - | 3 | 893 | |||
| AK148155.1 | - | 4 | 1033 | |||
| AK148326.1 | - | 3 | 886 | |||
| AK148383.1 | - | 3 | 886 | |||
| BC019991.1 | - | 3 | 888 | 4189407 | ||

Tspan10
Genomic Map