siRNA Details
| Gene Symbol: Tspan10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145363.2 | NP_663338.2 | 2 | 579 | ||
| GenBank mRNA | AK084214.1 | - | 2 | 526 | ||
| AK148037.1 | - | 2 | 522 | |||
| AK148155.1 | - | 3 | 662 | |||
| AK148326.1 | - | 2 | 515 | |||
| AK148383.1 | - | 2 | 515 | |||
| AK182989.1 | - | 1 | 344 | |||
| AK194285.1 | - | 1 | 341 | |||
| BC019991.1 | - | 2 | 517 | 4189407 | ||
| CK625019.1 | - | 2 | 579 | |||

Tspan10
Genomic Map